Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- H19
- Entrez Gene ID
-
100271857
- Gene Name
- H19, imprinted maternally expressed transcript (non-protein coding)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.12: 34334581 - 34336980 on Build EquCab3.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027326.3 |
|
2 - 3 |
1385 |
73 |
GenBank mRNA |
DN511008.1 |
- |
2 - 3 |
201 |
73 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC106781503 |
LOC106781503 |
|
-390.0
|
Anti-sense |
Chr. - 34334439 - 34334918 on Build EquCab3.0 |
Downstream |
microRNA mir-675 |
MIR675 |
eca-mir-675 |
315.0
|
Sense |
Chr. - 34335950 - 34336022 on Build EquCab3.0 |
View Full Product Details