Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IFT20
Entrez Gene ID
417671
Gene Name
intraflagellar transport 20
Gene Aliases
-
Chromosome Location
Chr.19: 9449067 - 9451237 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_415917.6 XP_415917.1 2 - 3 304 86
GenBank mRNA BX930995.2 - 2 - 3 267 86
CR352500.1 - 2 - 3 297 86

View Full Product Details