Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CABLES2
Entrez Gene ID
419229
Gene Name
Cdk5 and Abl enzyme substrate 2
Gene Aliases
-
Chromosome Location
Chr.20: 7938691 - 7957296 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015296655.2 XP_015152141.1 7 - 8 2247 75
XM_015296656.2 XP_015152142.1 6 - 7 2171 75
GenBank mRNA CR353110.1 - 5 - 6 525 75

View Full Product Details