Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM81A
Entrez Gene ID
415396
Gene Name
family with sequence similarity 81 member A
Gene Aliases
-
Chromosome Location
Chr.10: 7165593 - 7183550 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015278919.2 XP_015134405.1 8 - 9 3742 89
XM_015278920.2 XP_015134406.1 8 - 9 3726 89
XM_015278922.2 XP_015134408.1 7 - 8 3648 89
XM_015278924.2 XP_015134410.1 6 - 7 3580 89
XM_025154019.1 XP_025009787.1 8 - 9 3760 89
GenBank mRNA CR387250.1 - 2 - 3 235 89

View Full Product Details