Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBX1
Entrez Gene ID
418001
Gene Name
ring-box 1
Gene Aliases
-
Chromosome Location
Chr.1: 49860867 - 49871899 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199250.1 NP_001186179.1 3 - 4 298 78
XM_025148546.1 XP_025004314.1 3 - 4 371 78
GenBank mRNA CR390140.1 - 3 - 4 224 78

View Full Product Details