Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRR14L
Entrez Gene ID
416957
Gene Name
proline rich 14-like
Gene Aliases
C22orf30
Chromosome Location
Chr.15: 9177496 - 9196326 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015275419.2 XP_015130905.1 6 - 7 6577 100
XM_415250.6 XP_415250.4 6 - 7 6355 100
GenBank mRNA CR390992.1 - 3 - 4 642 100

View Full Product Details