Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MPHOSPH6
Entrez Gene ID
415811
Gene Name
M-phase phosphoprotein 6
Gene Aliases
-
Chromosome Location
Chr.11: 15802151 - 15809325 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277139.1 NP_001264068.1 4 - 5 446 122
GenBank mRNA BX929849.2 - 4 - 5 339 122
BX930843.2 - 4 - 5 355 122
BX935551.1 - 4 - 5 445 122
BX936086.2 - 4 - 5 414 122
CR406488.1 - 4 - 5 339 122

View Full Product Details