Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RUNX3
Entrez Gene ID
769687
Gene Name
runt related transcription factor 3
Gene Aliases
-
Chromosome Location
Chr.23: 2604900 - 2634767 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015297733.2 XP_015153219.1 3 - 4 802 80
GenBank mRNA EF051349.1 - 3 - 4 403 80

View Full Product Details