Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COLGALT1
Entrez Gene ID
424447
Gene Name
collagen beta(1-O)galactosyltransferase 1
Gene Aliases
COLGALT2, GLT25D2
Chromosome Location
Chr.8: 8109022 - 8142798 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015290549.2 XP_015146035.1 4 - 5 384 79
XM_422290.4 XP_422290.2 4 - 5 755 79
GenBank mRNA CR385470.1 - 4 - 5 456 79

View Full Product Details