Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IFT22
Entrez Gene ID
417503
Gene Name
intraflagellar transport 22
Gene Aliases
RABL5
Chromosome Location
Chr.19: 3655950 - 3659541 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_415752.6 XP_415752.2 3 - 4 248 95
GenBank mRNA CR406254.1 - 3 - 4 232 95
CR406564.1 - 2 - 3 223 95

View Full Product Details