Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIF2C
Entrez Gene ID
424583
Gene Name
kinesin family member 2C
Gene Aliases
-
Chromosome Location
Chr.8: 21097698 - 21113513 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015291291.2 XP_015146777.1 2 - 3 265 141
XM_015291292.2 XP_015146778.1 2 - 3 188 141
XM_015291293.2 XP_015146779.1 2 - 3 215 141
XM_015291294.2 XP_015146780.1 2 - 3 264 141
XM_015291295.2 XP_015146781.1 2 - 3 265 141
XM_015291297.2 XP_015146783.1 2 - 3 257 141
XM_015291299.2 XP_015146785.1 2 - 3 264 141
XM_025153279.1 XP_025009047.1 2 - 3 188 141
XM_422422.6 XP_422422.4 2 - 3 265 141
GenBank mRNA BX930594.2 - 2 - 3 226 141
BX931004.2 - 2 - 3 257 141
BX933284.2 - 2 - 3 164 141
CR391626.1 - 2 - 3 120 141
CR405925.1 - 2 - 3 207 141

View Full Product Details