Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CFAP221
Entrez Gene ID
424272
Gene Name
cilia and flagella associated protein 221
Gene Aliases
PCDP1
Chromosome Location
Chr.7: 28330064 - 28350684 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004942966.3 XP_004943023.1 19 - 20 1971 141
XM_422119.6 XP_422119.4 18 - 19 1946 141
GenBank mRNA BX950354.2 - 9 - 10 887 141

View Full Product Details