Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KMT2A
Entrez Gene ID
414897
Gene Name
lysine methyltransferase 2A
Gene Aliases
MLL
Chromosome Location
Chr.24: 4375990 - 4423673 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_025143342.1 XP_024999110.1 5 - 6 3519 68
XM_025143343.1 XP_024999111.1 5 - 6 3519 68
XM_025143344.1 XP_024999112.1 5 - 6 3519 68
XM_025143345.1 XP_024999113.1 6 - 7 2979 68
XM_025143346.1 XP_024999114.1 5 - 6 5889 68
GenBank mRNA AJ011002.1 - 4 - 5 3158 68

View Full Product Details