Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MARCH5
Entrez Gene ID
423815
Gene Name
membrane associated ring-CH-type finger 5
Gene Aliases
MARCH-V, MARCHF5, Mar-05
Chromosome Location
Chr.6: 21783365 - 21806151 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012906.1 NP_001012924.1 3 - 4 612 114
GenBank mRNA AJ720593.1 - 3 - 4 612 114
CR391018.1 - 3 - 4 429 114

View Full Product Details