Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDC2
Entrez Gene ID
374102
Gene Name
syndecan 2
Gene Aliases
syndecan-2
Chromosome Location
Chr.2: 126817624 - 126874833 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001462.1 NP_001001462.1 3 - 4 570 110
XM_025146997.1 XP_025002765.1 4 - 5 435 110
GenBank mRNA AB105811.1 - 3 - 4 321 110
AF508228.1 - 3 - 4 570 110
AF508229.1 - 3 - 4 570 110

View Full Product Details