Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
P3H1
Entrez Gene ID
414142
Gene Name
prolyl 3-hydroxylase 1
Gene Aliases
LEPRE1
Chromosome Location
Chr.21: 6377332 - 6383361 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001529.1 NP_001001529.1 7 - 8 1203 82
GenBank mRNA AY463528.1 - 7 - 8 1203 82

View Full Product Details