Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BPIFB2
Entrez Gene ID
395882
Gene Name
BPI fold containing family B member 2
Gene Aliases
BPIFB7, G2, TENP
Chromosome Location
Chr.20: 10296177 - 10301937 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205026.1 NP_990357.1 3 - 4 235 69
XM_015296335.2 XP_015151821.1 3 - 4 256 68
XM_025142176.1 XP_024997944.1 3 - 4 263 68
GenBank mRNA AB219157.1 - 2 - 3 190 68
AB219158.1 - 2 - 3 190 68
AB219159.1 - 2 - 3 190 68
AB219160.1 - 2 - 3 190 68
AF029841.1 - 3 - 4 235 69
HG007958.1 - 3 - 4 269 68
LC144559.1 - 2 - 3 190 68
LC144560.1 - 2 - 3 190 68
LC144561.1 - 2 - 3 190 68
LC144562.1 - 2 - 3 190 68
LC144563.1 - 2 - 3 190 68
LC144564.1 - 2 - 3 190 68
LC144565.1 - 2 - 3 190 68
LC144566.1 - 2 - 3 190 68
LC144567.1 - 2 - 3 190 68
LC144568.1 - 2 - 3 190 68
LC144569.1 - 2 - 3 190 68
LC144570.1 - 2 - 3 190 68
LC144571.1 - 2 - 3 190 68
LC144572.1 - 2 - 3 190 68
LC144573.1 - 2 - 3 190 68
LC144574.1 - 2 - 3 190 68
LC144575.1 - 2 - 3 190 68
LC144576.1 - 2 - 3 190 68
LC144577.1 - 2 - 3 190 68
LC144578.1 - 2 - 3 190 68
LC144579.1 - 2 - 3 190 68
LC144580.1 - 2 - 3 190 68
LC144581.1 - 2 - 3 190 68
LC144582.1 - 2 - 3 190 68
LC144583.1 - 2 - 3 190 68
LC144584.1 - 2 - 3 190 68
LC144585.1 - 2 - 3 190 68
LC144586.1 - 2 - 3 190 68
LC144587.1 - 2 - 3 190 68
LC144588.1 - 2 - 3 190 68
LC144589.1 - 2 - 3 190 68
LC144590.1 - 2 - 3 190 68
LC144591.1 - 2 - 3 190 68
LC144592.1 - 2 - 3 190 68
LC144593.1 - 2 - 3 190 68
LC144594.1 - 2 - 3 190 68
LC144595.1 - 2 - 3 190 68
LC144596.1 - 2 - 3 190 68
LC144597.1 - 2 - 3 190 68
LC144598.1 - 2 - 3 190 68
LC144599.1 - 2 - 3 190 68
LC144600.1 - 2 - 3 190 68
LC144601.1 - 2 - 3 190 68
LC144602.1 - 2 - 3 190 68
LC144603.1 - 2 - 3 190 68
LC144604.1 - 2 - 3 190 68
LC144605.1 - 2 - 3 190 68
LC144606.1 - 2 - 3 190 68
LC144607.1 - 2 - 3 190 68
LC144608.1 - 2 - 3 190 68

View Full Product Details