Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
B3GAT2
Entrez Gene ID
428638
Gene Name
beta-1,3-glucuronyltransferase 2
Gene Aliases
-
Chromosome Location
Chr.3: 82531592 - 82553687 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031527.1 NP_001026698.1 3 - 4 684 70
XM_025148810.1 XP_025004578.1 2 - 3 931 70
GenBank mRNA AJ889606.1 - 3 - 4 684 70

View Full Product Details