Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTBD1
Entrez Gene ID
415461
Gene Name
BTB domain containing 1
Gene Aliases
-
Chromosome Location
Chr.10: 11643177 - 11656460 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005819.1 NP_001005819.1 5 - 6 1119 108
GenBank mRNA AJ851743.1 - 5 - 6 1119 108
CR523906.1 - 4 - 5 637 108

View Full Product Details