Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB6A
Entrez Gene ID
419063
Gene Name
RAB6A, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.1: 196157062 - 196221090 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044640.1 NP_001038105.1 1 - 2 87 70
XM_015280973.2 XP_015136459.1 1 - 2 326 70
GenBank mRNA CR390639.1 - 1 - 2 313 70
DQ470138.1 - 1 - 2 87 70

View Full Product Details