Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAX3
Entrez Gene ID
374127
Gene Name
paired box 3
Gene Aliases
-
Chromosome Location
Chr.9: 8033232 - 8098638 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204269.1 NP_989600.1 6 - 7 979 77
XM_025153298.1 XP_025009066.1 6 - 7 1209 77
XM_025153299.1 XP_025009067.1 7 - 8 1555 77
GenBank mRNA AB080581.1 - 6 - 7 979 77

View Full Product Details