Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ICMT
Entrez Gene ID
419373
Gene Name
isoprenylcysteine carboxyl methyltransferase
Gene Aliases
-
Chromosome Location
Chr.21: 633594 - 636527 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006298.1 NP_001006298.1 1 - 2 256 72
GenBank mRNA AJ719287.1 - 1 - 2 256 72

View Full Product Details