Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NSA2
Entrez Gene ID
427206
Gene Name
NSA2, ribosome biogenesis homolog
Gene Aliases
TINP1
Chromosome Location
Chr.Z: 24202530 - 24206378 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006579.1 NP_001006579.1 2 - 3 215 123
GenBank mRNA AJ719405.1 - 2 - 3 215 123
CR388679.1 - 2 - 3 257 123
CR523694.1 - 3 - 4 128 123

View Full Product Details