Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMOD4
Entrez Gene ID
395547
Gene Name
tropomodulin 4
Gene Aliases
SK-TMOD, tropomodulin-4
Chromosome Location
Chr.25: 253777 - 257285 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204774.1 NP_990105.1 8 - 8 851 100
GenBank mRNA AB052717.1 - 7 - 7 773 100
AF165215.1 - 8 - 8 851 100

View Full Product Details