Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SS
Entrez Gene ID
547508
Gene Name
sucrose synthase
Gene Aliases
N-100
Chromosome Location
Chr.13: 21211899 - 21217220 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001250596.1 NP_001237525.1 12 - 13 2460 86
GenBank mRNA AF030231.1 - 12 - 13 2460 86
AK285864.1 - 12 - 13 2471 86

View Full Product Details