Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SS
Entrez Gene ID
547508
Gene Name
sucrose synthase
Gene Aliases
N-100
Chromosome Location
Chr.13: 21211899 - 21217220 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001250596.1 NP_001237525.1 5 - 6 641 64
GenBank mRNA AF030231.1 - 5 - 6 641 64
AK285864.1 - 5 - 6 652 64

Gene Symbol
LOC100788584
Entrez Gene ID
100788584
Gene Name
sucrose synthase-like
Gene Aliases
GmSS
Chromosome Location
Chr.17: 3412753 - 3418155 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK285909.1 - 5 - 6 651 64

View Full Product Details