Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GDH2
Entrez Gene ID
547688
Gene Name
glutamate dehydrogenase 2
Gene Aliases
-
Chromosome Location
Chr.17: 12459358 - 12463310 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001249488.1 NP_001236417.1 7 - 8 1058 97
GenBank mRNA AJ879896.1 - 7 - 8 1058 97

View Full Product Details