Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY38
Entrez Gene ID
100170677
Gene Name
transcription factor
Gene Aliases
-
Chromosome Location
Chr.18: 57861351 - 57870248 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001250616.1 NP_001237545.1 1 - 2 334 70
GenBank mRNA EU375347.1 - 1 - 2 334 70

View Full Product Details