Gene Transcripts

Gene Symbol
PTTG1IP
Entrez Gene ID
754
Gene Name
pituitary tumor-transforming 1 interacting protein
Gene Aliases
C21orf1, C21orf3, PBF
Chromosome Location
Chr. 21: 44849585 - 44873903 on Build GRCh38
UniGene ID
Hs.474010
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004339.3 NP_004330.1 2, 3 447
GenBank mRNA AF149785.1 - 2, 3 385
AK094882.1 - 2, 3 180
AK290139.1 - 2, 3 200
AK315626.1 - 2, 3 262
BC000415.2 - 2, 3 204 2961420
BC012858.1 - 2, 3 230 3857505
BC019295.1 - 2, 3 194 2823814
BC020983.1 - 2, 3 211 3851447
BC031097.1 - 2, 3 225 5288388
BC034250.1 - 2, 3 219 3862830
BG721942.1 - 2, 3 386
DC396020.1 - 2, 3 448
Z50022.1 - 2, 3 253

View Full Product Details