Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001972.3 | NP_001963.1 | 3 | 468 | |
XM_011527775.1 | XP_011526077.1 | 4 | 1145 | ||
XM_011527776.1 | XP_011526078.1 | 4 | 484 | ||
GenBank mRNA | BC074816.2 | - | 3 | 385 | |
BC074817.2 | - | 3 | 385 | 30915320 | |
D00187.1 | - | 2 | 258 | ||
J03545.1 | - | 1 | 126 | ||
M34379.1 | - | 3 | 386 | ||
X05875.1 | - | 3 | 386 |