Gene Transcripts

Gene Symbol
IL3
Entrez Gene ID
3562
Gene Name
interleukin 3
Gene Aliases
IL-3, MCGF, MULTI-CSF
Chromosome Location
Chr. 5: 132060654 - 132063203 on Build GRCh38
UniGene ID
Hs.694
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000588.3 NP_000579.2 3 305
GenBank mRNA BC066272.1 - 3 298
BC066273.1 - 3 298 6971774
BC066274.1 - 3 298 6971775
BC066275.1 - 3 298 6971776
BC066276.1 - 3 298 6971777
BC069472.1 - 3 301 7262122
M14743.1 - 3 261
M17115.1 - 3 290
M20137.1 - 3 305

View Full Product Details