Gene Transcripts

Gene Symbol
OR1D2
Entrez Gene ID
4991
Gene Name
olfactory receptor family 1 subfamily D member 2
Gene Aliases
OLFR1, OR17-4
Chromosome Location
Chr. 17: 3092058 - 3092996 on Build GRCh38
UniGene ID
Hs.532771
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002548.2 NP_002539.2 1 414
GenBank mRNA BC069552.1 - 1 414
BC106735.1 - 1 414 40015766
BC106736.1 - 1 414 40015767

View Full Product Details