Gene Transcripts
- Gene Symbol
- PAFAH1B1
- Entrez Gene ID
- 5048
- Gene Name
- platelet activating factor acetylhydrolase 1b regulatory subunit 1
- Gene Aliases
- LIS1, LIS2, MDCR, MDS, NudF, PAFAH
- Chromosome Location
- Chr. 17: 2593210 - 2685617 on Build GRCh38
- UniGene ID
- Hs.77318
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000430.3 |
NP_000421.1
|
8, 9 |
1460 |
|
XM_011523901.1 |
XP_011522203.1
|
9, 10 |
1509 |
|
XM_011523902.2 |
XP_011522204.1
|
9, 10 |
1341 |
|
XM_011523903.2 |
XP_011522205.1
|
9, 10 |
1259 |
|
XM_017024699.1 |
XP_016880188.1
|
8, 9 |
1210 |
|
XM_017024700.1 |
XP_016880189.1
|
8, 9 |
1287 |
|
XM_017024701.1 |
XP_016880190.1
|
8, 9 |
1203 |
|
XM_017024702.1 |
XP_016880191.1
|
7, 8 |
1048 |
|
GenBank mRNA |
AF208835.1 |
- |
7, 8 |
893 |
|
AF208836.1 |
- |
7, 8 |
893 |
|
AF208837.1 |
- |
7, 8 |
892 |
|
AF208838.1 |
- |
7, 8 |
892 |
|
AF400434.1 |
- |
7, 8 |
1085 |
|
AK303015.1 |
- |
4, 5 |
700 |
|
AK313078.1 |
- |
8, 9 |
1114 |
|
BC064638.1 |
- |
8, 9 |
1102 |
5786560 |
L13385.1 |
- |
8, 9 |
1109 |
|
L13386.1 |
- |
8, 9 |
1345 |
|
L13387.1 |
- |
|
1328 |
|
L25107.1 |
- |
|
540 |
|
S81396.1 |
- |
4, 5 |
538 |
|
View Full Product Details