Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006219.2 | NP_006210.1 | 1 | 51 | |
XM_005247530.2 | XP_005247587.1 | 3 | 400 | ||
XM_006713659.3 | XP_006713722.1 | 2 | 271 | ||
XM_011512895.2 | XP_011511197.1 | 4 | 396 | ||
XM_017006619.1 | XP_016862108.1 | 2 | 74 | ||
GenBank mRNA | BC114432.1 | - | 1 | 39 | |
S67334.1 | - | 1 | 35 |