Gene Transcripts

Gene Symbol
POU2F1
Entrez Gene ID
5451
Gene Name
POU class 2 homeobox 1
Gene Aliases
OCT1, OTF1, oct-1B
Chromosome Location
Chr. 1: 167220829 - 167427345 on Build GRCh38
UniGene ID
Hs.283402
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198783.1 NP_001185712.1 9 1162
NM_001198786.1 NP_001185715.1 8 840
NM_002697.3 NP_002688.3 9 1029
NR_037163.1 10 1073
XM_011509653.2 XP_011507955.1 10 981
XM_011509654.2 XP_011507956.1 9 942
XM_011509655.1 XP_011507957.1 10 981
XM_017001508.1 XP_016856997.1 10 981
GenBank mRNA AK091438.1 - 11 1457
AK302525.1 - 11 1135
AK303201.1 - 8 778
AK308004.1 - 10 1068
AY113189.1 - 9 981
BC001664.2 - 9 945 2966289
BC003571.1 - 9 945 3544378
BC007388.2 - 5 615
BC052274.1 - 10 1053 4622256
KT438684.1 - 11 1140
X13403.1 - 9 950

View Full Product Details