Gene Transcripts
- Gene Symbol
- QDPR
- Entrez Gene ID
- 5860
- Gene Name
- quinoid dihydropteridine reductase
- Gene Aliases
- DHPR, PKU2, SDR33C1
- Chromosome Location
- Chr. 4: 17486393 - 17512234 on Build GRCh38
- UniGene ID
- Hs.75438
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000320.2 |
NP_000311.2
|
6, 7 |
797 |
|
NM_001306140.1 |
NP_001293069.1
|
5, 6 |
704 |
|
GenBank mRNA |
AK124382.1 |
- |
5, 6 |
556 |
|
AK223437.1 |
- |
6, 7 |
653 |
|
AK289773.1 |
- |
6, 7 |
653 |
|
AK296622.1 |
- |
5, 6 |
544 |
|
BC000576.2 |
- |
6, 7 |
641 |
3162690 |
BM762805.1 |
- |
4, 5 |
595 |
|
M16447.1 |
- |
6, 7 |
697 |
|
X04882.1 |
- |
6, 7 |
641 |
|
View Full Product Details