Gene Transcripts

Gene Symbol
RDX
Entrez Gene ID
5962
Gene Name
radixin
Gene Aliases
DFNB24
Chromosome Location
Chr. 11: 110174880 - 110296712 on Build GRCh38
UniGene ID
Hs.263671
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001260492.1 NP_001247421.1 5 576
NM_001260493.1 NP_001247422.1 5 576
NM_001260496.1 NP_001247425.1 4 480
NM_002906.3 NP_002897.1 5 576
GenBank mRNA AK312903.1 - 5 478
BC002626.2 - 5 460
BC020751.1 - 5 476
BC047109.1 - 5 476 5284438
BI600187.1 - 5 587
DQ916738.1 - 5 576
DQ916739.1 - 4 480
DQ916741.1 - 5 576
DQ916742.1 - 5 576
L02320.1 - 4 296

View Full Product Details