Gene Transcripts

Gene Symbol
CXCL5
Entrez Gene ID
6374
Gene Name
C-X-C motif chemokine ligand 5
Gene Aliases
ENA-78, SCYB5
Chromosome Location
Chr. 4: 73995642 - 73998729 on Build GRCh38
UniGene ID
Hs.89714
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002994.4 NP_002985.1 3 407
GenBank mRNA AK312128.1 - 3 355
BC008376.1 - 3 358 3826939
BT006807.1 - 3 259
BU169770.1 - 3 378
CR457428.1 - 3 259
DN992282.1 - 3 465
X78686.1 - 3 365

View Full Product Details