Gene Transcripts

Gene Symbol
SNRPA
Entrez Gene ID
6626
Gene Name
small nuclear ribonucleoprotein polypeptide A
Gene Aliases
Mud1, U1-A, U1A
Chromosome Location
Chr. 19: 40750854 - 40765392 on Build GRCh38
UniGene ID
Hs.466775
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004596.4 NP_004587.1 4, 5 1162
GenBank mRNA AK293140.1 - 3, 4 558
AK313182.1 - 4, 5 775
BC000405.2 - 4, 5 1142 2822987
BC008290.1 - 4, 5 726 3677365
BQ002918.1 - 2, 3 480
X06347.1 - 4, 5 732

View Full Product Details