Gene Transcripts
- Gene Symbol
- SPAST
- Entrez Gene ID
- 6683
- Gene Name
- spastin
- Gene Aliases
- ADPSP, FSP2, SPG4
- Chromosome Location
- Chr. 2: 32063551 - 32157637 on Build GRCh38
- UniGene ID
- Hs.468091
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014946.3 |
NP_055761.2
|
16, 17 |
1952 |
|
NM_199436.1 |
NP_955468.1
|
15, 16 |
1856 |
|
XM_005264516.4 |
XP_005264573.1
|
16, 17 |
1728 |
|
XM_011533067.2 |
XP_011531369.1
|
15, 16 |
1941 |
|
XM_017004777.1 |
XP_016860266.1
|
15, 16 |
1913 |
|
XM_017004778.1 |
XP_016860267.1
|
14, 15 |
1845 |
|
GenBank mRNA |
AB029006.1 |
- |
15, 16 |
1856 |
|
AJ246001.1 |
- |
16, 17 |
1856 |
|
AK304135.1 |
- |
7, 8 |
466 |
|
BC150260.1 |
- |
15, 16 |
1856 |
9007202 |
View Full Product Details