Gene Transcripts

Gene Symbol
SUPT6H
Entrez Gene ID
6830
Gene Name
SPT6 homolog, histone chaperone
Gene Aliases
SPT6, SPT6H, emb-5
Chromosome Location
Chr. 17: 28662183 - 28702684 on Build GRCh38
UniGene ID
Hs.250429
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320755.1 NP_001307684.1 25, 26 3417
NM_003170.4 NP_003161.2 24, 25 3315
XM_017024978.1 XP_016880467.1 26, 27 3476
XM_017024979.1 XP_016880468.1 24, 25 3553
XM_017024980.1 XP_016880469.1 23, 24 3247
GenBank mRNA AL833607.1 - 25, 26 3418
BC003696.1 - 9, 10 1190
BC017105.1 - 4, 5 458
BC136522.1 - 24, 25 3227 9020512
BC136524.1 - 24, 25 3227 9020514
BC150268.1 - 24, 25 3229 9007230
D79984.1 - 24, 25 3229

View Full Product Details