Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001202476.1 | NP_001189405.1 | 3 | 155 | |
NM_003341.4 | NP_003332.1 | 4 | 450 | ||
NM_182666.2 | NP_872607.1 | 3 | 399 | ||
XM_005265431.4 | XP_005265488.1 | 3 | 207 | ||
GenBank mRNA | AK295354.1 | - | 3 | 155 | |
AK314854.1 | - | 4 | 359 | ||
BC009139.1 | - | 4 | 374 | 3853408 | |
BI223271.1 | - | 3 | 294 | ||
DB478686.1 | - | 4 | 454 | ||
DB499676.1 | - | 3 | 155 | ||
X92963.1 | - | 3 | 221 |