Gene Transcripts
- Gene Symbol
- UBE2H
- Entrez Gene ID
- 7328
- Gene Name
- ubiquitin conjugating enzyme E2 H
- Gene Aliases
- E2-20K, GID3, UBC8, UBCH, UBCH2
- Chromosome Location
- Chr. 7: 129830732 - 129952960 on Build GRCh38
- UniGene ID
- Hs.643548
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001202498.1 |
NP_001189427.1
|
2, 3 |
146 |
|
NM_003344.3 |
NP_003335.1
|
2, 3 |
542 |
|
NM_182697.2 |
NP_874356.1
|
2, 3 |
542 |
|
GenBank mRNA |
AB451490.1 |
- |
2, 3 |
137 |
|
AK316590.1 |
- |
2, 3 |
125 |
|
AY302138.1 |
- |
2, 3 |
137 |
|
BC006277.2 |
- |
2, 3 |
454 |
3957107 |
BI088146.1 |
- |
2, 3 |
146 |
|
BT006756.1 |
- |
2, 3 |
137 |
|
CB995681.1 |
- |
|
559 |
|
KJ534959.1 |
- |
2, 3 |
137 |
|
Z29328.1 |
- |
2, 3 |
190 |
|
Z29330.1 |
- |
2, 3 |
190 |
|
View Full Product Details