Gene Transcripts

Gene Symbol
UPK2
Entrez Gene ID
7379
Gene Name
uroplakin 2
Gene Aliases
UP2, UPII
Chromosome Location
Chr. 11: 118956298 - 118958559 on Build GRCh38
UniGene ID
Hs.379613
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006760.3 NP_006751.1 3, 4 407
GenBank mRNA AY455312.1 - 3, 4 354
BC113900.1 - 3, 4 385 40029623
Y13645.1 - 3, 4 392

View Full Product Details