Gene Transcripts
- Gene Symbol
- SPOP
- Entrez Gene ID
- 8405
- Gene Name
- speckle type BTB/POZ protein
- Gene Aliases
- BTBD32, TEF2
- Chromosome Location
- Chr. 17: 49598884 - 49678163 on Build GRCh38
- UniGene ID
- Hs.463382
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001007226.1 |
NP_001007227.1
|
4, 5 |
552 |
|
NM_001007227.1 |
NP_001007228.1
|
3, 4 |
430 |
|
NM_001007228.1 |
NP_001007229.1
|
2, 3 |
378 |
|
NM_001007229.1 |
NP_001007230.1
|
3, 4 |
429 |
|
NM_001007230.1 |
NP_001007231.1
|
4, 5 |
481 |
|
NM_003563.3 |
NP_003554.1
|
3, 4 |
500 |
|
XM_005257723.4 |
XP_005257780.1
|
3, 4 |
724 |
|
XM_005257724.4 |
XP_005257781.1
|
2, 3 |
654 |
|
XM_017025204.1 |
XP_016880693.1
|
1, 2 |
167 |
|
GenBank mRNA |
AJ000644.1 |
- |
2, 3 |
238 |
|
AK222589.1 |
- |
2, 3 |
203 |
|
AK293045.1 |
- |
3, 4 |
711 |
|
AK293587.1 |
- |
2, 3 |
320 |
|
AK312691.1 |
- |
2, 3 |
214 |
|
BC001269.1 |
- |
3, 4 |
384 |
3461565 |
BC003385.1 |
- |
2, 3 |
195 |
3445878 |
CB992730.1 |
- |
4, 5 |
453 |
|
View Full Product Details