Gene Transcripts

Gene Symbol
NR0B2
Entrez Gene ID
8431
Gene Name
nuclear receptor subfamily 0 group B member 2
Gene Aliases
SHP, SHP1
Chromosome Location
Chr. 1: 26911484 - 26914076 on Build GRCh38
UniGene ID
Hs.427055
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021969.2 NP_068804.1 2 844
GenBank mRNA BC030207.1 - 2 734
HQ692833.1 - 2 708

View Full Product Details