Gene Transcripts
- Gene Symbol
- LAMTOR3
- Entrez Gene ID
- 8649
- Gene Name
- late endosomal/lysosomal adaptor, MAPK and MTOR activator 3
- Gene Aliases
- MAP2K1IP1, MAPBP, MAPKSP1, MP1, PRO0633, Ragulator3
- Chromosome Location
- Chr. 4: 99878338 - 99894546 on Build GRCh38
- UniGene ID
- Hs.433332
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001243736.1 |
NP_001230665.1
|
6, 7 |
536 |
|
NM_021970.3 |
NP_068805.1
|
6, 7 |
557 |
|
NR_024170.1 |
|
5, 6 |
511 |
|
GenBank mRNA |
AF130115.1 |
- |
5, 6 |
357 |
|
AF201947.1 |
- |
6, 7 |
467 |
|
AK223311.1 |
- |
6, 7 |
448 |
|
AK311755.1 |
- |
6, 7 |
418 |
|
AL544981.3 |
- |
6, 7 |
366 |
|
BC026245.1 |
- |
6, 7 |
450 |
4808855 |
BC110402.1 |
- |
5, 6 |
377 |
|
BI562630.1 |
- |
5, 6 |
514 |
|
View Full Product Details