Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003906.4 | NP_003897.2 | 10 | 2791 | |
XM_005261203.4 | XP_005261260.1 | 11 | 3367 | ||
XM_005261204.4 | XP_005261261.1 | 11 | 3263 | ||
XM_005261205.3 | XP_005261262.1 | 11 | 2879 | ||
GenBank mRNA | AB011144.1 | - | 10 | 2361 | |
AJ010089.1 | - | 10 | 2720 | ||
AK126313.1 | - | 1 | 973 | ||
AK304208.1 | - | 10 | 1650 | ||
BC104958.1 | - | 10 | 2759 | 8143961 | |
BC104960.1 | - | 10 | 2759 | 8143963 |