Gene Transcripts
- Gene Symbol
- NEMF
- Entrez Gene ID
- 9147
- Gene Name
- nuclear export mediator factor
- Gene Aliases
- NY-CO-1, SDCCAG1
- Chromosome Location
- Chr. 14: 49782083 - 49853073 on Build GRCh38
- UniGene ID
- Hs.655964
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_017021760.1 |
XP_016877249.1
|
31, 32 |
3146 |
|
XM_017021761.1 |
XP_016877250.1
|
26, 27 |
2599 |
|
GenBank mRNA |
AF039687.1 |
- |
10, 11 |
1173 |
|
AK093783.1 |
- |
16, 17 |
1583 |
|
AK304695.1 |
- |
31, 32 |
3116 |
|
AK314281.1 |
- |
14, 15 |
1322 |
|
BC020794.2 |
- |
12, 13 |
1237 |
|
BC064364.1 |
- |
12, 13 |
1229 |
|
BC107764.1 |
- |
13, 14 |
1297 |
|
BX640804.1 |
- |
8, 9 |
802 |
|
BX640807.1 |
- |
15, 16 |
1439 |
|
BX648598.1 |
- |
23, 24 |
2687 |
|
View Full Product Details