Gene Transcripts
- Gene Symbol
- PSMD6
- Entrez Gene ID
- 9861
- Gene Name
- proteasome 26S subunit, non-ATPase 6
- Gene Aliases
- Rpn7, S10, SGA-113M, p42A, p44S10
- Chromosome Location
- Chr. 3: 64010549 - 64024010 on Build GRCh38
- UniGene ID
- Hs.152536
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271779.1 |
NP_001258708.1
|
5, 6 |
1073 |
|
NM_001271780.1 |
NP_001258709.1
|
4, 5 |
788 |
|
NM_001271781.1 |
NP_001258710.1
|
4, 5 |
663 |
|
NM_014814.2 |
NP_055629.1
|
4, 5 |
914 |
|
XM_005265619.1 |
XP_005265676.1
|
4, 5 |
1010 |
|
XM_011534288.1 |
XP_011532590.1
|
5, 6 |
969 |
|
XM_017007569.1 |
XP_016863058.1
|
5, 6 |
816 |
|
GenBank mRNA |
AF215935.1 |
- |
4, 5 |
780 |
|
AF530062.1 |
- |
4, 5 |
727 |
|
AK095059.1 |
- |
3, 4 |
1496 |
|
AK290205.1 |
- |
4, 5 |
768 |
|
AL529900.3 |
- |
4, 5 |
649 |
|
AY359879.1 |
- |
4, 5 |
760 |
|
AY568086.1 |
- |
4, 5 |
752 |
|
BC000630.1 |
- |
4, 5 |
744 |
3344973 |
BC000904.2 |
- |
4, 5 |
765 |
2900971 |
BC012369.1 |
- |
4, 5 |
762 |
3858094 |
BX647742.1 |
- |
5, 6 |
940 |
|
D14663.1 |
- |
4, 5 |
752 |
|
View Full Product Details